Learn More
Honeywell Analytics™ Biosystems PhD6 Multigas Detector, O2, CO, H2S, LEL
Supplier: Honeywell Analytics™ 545490
Description
Oxygen (O2); Range: 0 to 30%/volume; Resolution: 0.1%
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Oxygen | |
0.1% | |
-20°C to +50°C |
0 to 30% | |
-4°F to +122°F |