Learn More
Azzota Corp Holmium Filter Spectrophotometer Calibration Tools Use For Testing Spectrophotometer Wavelength Accuracy Calibrated From 200 To 700 Nm Accuracy +/- 0.2Nm Filter Size 10 X 10 X 45 mm
Supplier:  Azzota Corp Q802
Encompass_Preferred
Holmium Filter Spectrophotometer calibration tools Use for testing spectrophotometer wavelength accuracy Calibrated from 200 to 700 nm Accuracy +/- 0.2nm Filter size 10 x 10 x 45 mm
Catalog No. 50-753-2895