Learn More
Electron Microscopy Sciences Hollande's Fixative Prefilled, 60 ml cup, 30 ml Fill 30 ML
Supplier: Electron Microscopy Sciences 6412030
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A modification of Bouin's solution. It is stable and will decalcify small bone specimens. Tissue that is fixed with Hollande's can be stained successfully with most stains, and the cupric acetate in the solution stabilizes red blood cell membranes and cosinophil and endocrine cell granules so that less lysis occurs than with Bouin's solution. Hollande's is widely used as a fixative for biopsy specimens of the gastrointestinal tract.
Thorough washing of the fixative prior to placing the specimen in a phosphate buffered formalin solution is necessary because the salts present in the solution will form an insoluble phosphate precipitate. Packaged in leak-proof histo-containers; prefilled ready-to-use.
Cup: 60 ml
Fill: 30 ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.