Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 4-6 nm thick on 200 mesh Gold Grid
Supplier: Electron Microscopy Sciences HC200AUUL
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Gold
Thickness: 4-6 nm