Learn More
Glas Col LLC Shaker Accessory
Supplier: Glas Col LLC 099A MFG3
This tube holder lets you use our Bench Top Shaker for Mojonnier fat extraction. Each holder accepts up to four flasks, 37mm in diameter, with a load capacity of up to 5 pounds.