Learn More
Zymo Research Corporation Human HCT116 DKO Non-Methylated DNA (5 μg / 20 μl)
Supplier: Zymo Research Corporation D50141
The Human HCT116 DKO Non-methylated DNA is purified from cells that contain genetic knockouts of both DNA methyltransferases DNMT1 (-/-) and DNMT3b (-/-).The DNA derived from HCT116 DKO cells has a low level of DNA methylation and can be used as a control for DNA methylation analysis. The Human HCT116 DKO Methylated DNA is purified HCT116 DKO DNA that has been enzymatically methylated at all cytosine positions comprising CG dinucleotides by M.SssI methyltransferase2 (EC 2.1.1.37) and can be used as a positive control for DNA methylation analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.