Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ HM 650V Vibrating Blade Microtome Accessories
$1348.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
41-915-0
|
Epredia™
419150 |
Microtome Blade Holder |
Each for $1,348.00
|
|
|||||
52-501-3
|
Epredia™
525013 |
PTU-3 Pump and Tank | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Standard buffer tray
- Holds stages in place using a magnet
- Fill tray with all water-based buffers and solutions
- Use pre-cooled in the standard buffer tray for cooling tissue
- Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
- Magnification 2.5X
- Move, swivel, and displace for optimal viewing position
- Includes adapter and illumination
- Eyepieces: 10X
- Supplementary lens: 0.63X
- Zoom magnifying range: 4.1 to 31.5X
- Microscope adapter for angled view of specimen
- Horizontal view and vertical drive and swivel mechanism
- 3.5mm fiber bundle
- Swan-neck light guide: 50mm long
- Flexible light guide: 750mm long
- High light intensity
- Include adapter for the KL200 (760317)
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title