Learn More
Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories
Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.
Supplier: Epredia™ 705200
Description
Standard buffer tray
Cooling element for buffer solution
Large-field magnifier
StemiScope™ with optics and mount
Swan-neck light guide
Specifications
Blade Holder (for insertion of low-profile blades) | |
Sliding Microtome Accessories | |
Epredia HM 430 and HM 450 Sliding Microtomes |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov