Learn More
Simport™ Scientific Histosette™ I Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ I Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
$424.00 - $438.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-045-044
|
Simport™ Scientific
M4902 |
White |
Case of 1500 for $438.00
|
|
|||||
22-045-045
|
Simport™ Scientific
M4903 |
Pink |
Case of 1500 for $438.00
|
|
|||||
22-045-046
|
Simport™ Scientific
M4904 |
Green |
Case of 1500 for $438.00
|
|
|||||
22-045-047
|
Simport™ Scientific
M4905 |
Yellow |
Case of 1500 for $438.00
|
|
|||||
22-045-048
|
Simport™ Scientific
M4906 |
Blue |
Case of 1500 for $438.00
|
|
|||||
22-045-049
|
Simport™ Scientific
M4907 |
Peach |
Case of 1500 for $424.00
|
|
|||||
22-045-050
|
Simport™ Scientific
M4908 |
Tan |
Case of 1500 for $424.00
|
|
|||||
22-045-051
|
Simport™ Scientific
M4909 |
Gray |
Case of 1500 for $438.00
|
|
|||||
22-045-052
|
Simport™ Scientific
M49010 |
Lilac |
Case of 1500 for $438.00
|
|
|||||
22-045-053
|
Simport™ Scientific
M49011 |
Orange |
Case of 1500 for $438.00
|
|
|||||
22-045-054
|
Simport™ Scientific
M49012 |
Aqua |
Case of 1500 for $438.00
|
|
|||||
Description
- Made of acetal
- Keeps specimens safely submerged in liquid
- Resistant to the chemical action of most histological solvents
- 30° angle anterior writing area
- One-piece construction with frangible hinges for easy lid removal
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Yes | |
Acetal | |
Tissue Cassette |
For in vitro diagnostic (IVD) use only.