Learn More
Electron Microscopy Sciences Histology Control Microscope Slide 334+
SDP
Supplier:  Electron Microscopy Sciences 6344820
Encompass_Preferred
Use our Histology Control Slides and you will always know for sure which specimen is the Patients and which is the Control. Style 334+ has 1 inch frosted area.
Catalog No. 50-251-13