Learn More
Revvity Health Sciences Inc Histamine Dynamic Acylation Reagent Diluent, 1 Item
Supplier:  Revvity Health Sciences Inc 62HTMDDDA
Encompass
The Histamine dynamic acylation reagent spare is a mandatory prerequisite treatment for the quantification of histamine.
Catalog No. 50-211-9120