Learn More
CTL Scientific Supply Corp HISOCOLOR ECO SULPHITE
SDP
Supplier:  CTL Scientific Supply Corp 931095
Encompass_Preferred
Environmental test kit for Sulphite Gradation of scale: 1 drop corresponds to 1mg/l 60 tests/kit
Catalog No. 50-300-386