Learn More
Cytiva HiScale 10/40 column
Supplier: Cytiva 29360550
HiScale are easy to pack empty columns designed for medium to high pressure chromatography up to 20 bar (290 psi) using a chromatography system. 10mm x 400mm