Learn More
Genscript Corporation Antibody Kit Protein Purification Protein Purification 1/EA
Supplier:  Genscript Corporation L00440B
Encompass_Preferred
His Tag Antibody Plate is a microtiter plate coated with THETM Anti His Tag Antibody. The plate is designed for capturing His tagged protein with His tag located at N-terminus C-terminus or internal of the protein. It can be applied for analysis of different length of His tag including Hexa-His (6 His) Penta-His (5-His) and Tetra-His (4-His).The plate demonstrate high sensitivity and high binding capacity. The capacity and sensitivity varies depending on protein size structure and solution buffer. Generally proteins with lower molecular weight (M.W) can be more sensitive to the His antibody plate than ones with higher M.W. Compared with Nickel Coated Plate the affinity plate can bind target proteins with high specificity and tolerable with several common used reagents such as EDTA imidazole andBeta-ME.
Catalog No. 50-252-9994