Learn More
Cytiva His Capture Kit, type 2
Supplier: Cytiva 29234602
Reagents for capture of histidine-tagged molecules as ligands in biomolecular interaction analysis. Designed for use with Biacore 8K (larger kit component volumes than in His Capture Kit) but can be used with all other Biacore systems.