Learn More
Cole-Parmer Stuart SMP20 High Resolution Digital Melting Point Apparatus, 230 V
Supplier: Cole-Parmer 0301147
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens with built-in illumination. Extend the back feet for the optimal viewing angle.
Set a plateau temperature for initial warm-up — unattended warm-up time frees up the user until the plateau temperature is reached. Heating rate continues at 2°C after the warm-up, slowly reaching the melt temperature for greater accuracy.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.