Learn More
Fine Science Tools Hippocampal Tool, Tip: 8.5 x 14.25 x 0.3 mm, Stainless Steel, 16 cm
Supplier: Fine Science Tools 1009915
Individual Item Name: Hippocampal Tool - 16cm/ Tip Shape: Straight/ Tip Dimensions: 8.5 x 14.25 x 0.3mm/ Alloy: Stainless Steel/ Length: 16cm