Learn More
Cole-Parmer Oakton pH Probe, Sealed, DJ, Epoxy, High pH Sodium, 3 ft Cable, BNC
Supplier:  Cole-Parmer 3580506
Encompass_Preferred
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.
Catalog No. NC9654617