Learn More
Showa Denko America Inc HILICpak VG-50 2D
Supplier: Showa Denko America Inc F7630300
Suitable for saccharides and polar sample analysis using HILIC mode, very high recovery rate of reducing saccharides vs other polymer/silica based columns, specially suited for LC/MS, dimensions: 2.0 mm ID x 150 mm L