Learn More
Showa Denko America Inc HILICpak VC-50 2D
Supplier:  Showa Denko America Inc F7630700
Encompass_Preferred
Modified carboxyl groups is suitable for cationic substance analysis including amines, amino acids and sugars, dominant separation mode by RP using HILIC mode as secondary interaction, this least polar column of HILICpak polymer-based packing material provides stability and minimal deterioration over time, dimensions: 2 mm ID x 150 mm L
Catalog No. 50-161-3266