Learn More
OHAUS™ High Speed Microplate Shaker, SHHSMPDG
High Speed Microplate Shaker provides variable high speed shaking from 600 to 2500 rpm for microplates, deep well plates, and small-diameter tubes up to 5 in. (12.7 cm) tall.
Supplier:  OHAUS™ 30573784
Catalog No. 02-217-061
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Sturdy, adjustable top plate secures up to 48 microplates, 6 deep well plates, or small tubes less than 5 in. (12.7 cm) tall and up to 7 lb. (3.2 kg).
  • For difficult to mix samples, or to prevent overheating samples while shaking, the Pulse Feature can be adjusted to Pulse ON and Pulse OFF for intervals of between 1 and 59 seconds.
  • Independent LEDs and Touchpad Control for Speed and Time allow you to accurately reproduce the exact settings every time.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
600 rpm to 2500 rpm
48 Microplates
311 mm
279 mm
SHHSMPDG
120 V
12.25 in.
11 in.
Digital
Product Suggestions
Product Suggestions