Learn More
Agilent Technologies Fragment Analyzer DNA/NGS Kits
Supplier: Agilent Technologies DNF4741000
HS NGS Fragment Kit (1-6000 bp) - Fragment Analyzer system kit for the quantitative and qualitative analysis of NGS libraries and intermediates from 100 to 6,000 bp at low concentrations. Incudes the reagents necessary for running 1,000 samples.