Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ High Precision Dressing Forceps with Serrated Tips
Used for holding dressing materials during procedures. Fisherbrand High Precision Dressing Forceps with Serrated Tips are thumb forceps and are made of stainless steel.
This item is not returnable.
View return policy
Description
- Straight design with serrated tips
- Replacement for PL-DIS-2
Specifications
Serrated | |
4.25 in. | |
110 mm | |
High |
Straight | |
Stainless Steel | |
Forceps, Dressing | |
Tip: 0.5 x 0.75 mm (0.020 x 0.030 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction