Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ High-Power Electronic Crimping Tool with OLED Display Screen

Fixed power supply provides steady, reliable power; Cords are available for China, Europe, Japan, Taiwan, United Kingdom, and United States.
Supplier: Restek™ 23236CN
Description
- Buy only the size crimper or decapper needed (11 mm or 20 mm) with interchangable jaw sets, sold separately
- The separate crimper and decapper jaw sets fit standard lab vial seals that are aluminum or two-part aluminum/steel caps
- Ergonomic design reduces strain and arm injury associated with many manual crimping tools
- Optimize crimping power for each cap type, monitor battery use, and view lifetime statistics with the crisp OLED screen display
- Save time and minimize adjustments by storing separate programs for different caps and seals
- Strong enough for use with all steel and magnetic caps
- Combine with optional base and mounting kit for hands-free operation
- Two convenient sleep mode options
Specifications
Crimping Tool | |
11 mm or 20 mm | |
1 Ea. |
Crimp Caps | |
Universal Fixed Power Supply, USB Drive | |
Power Cord (China) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title