Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSC Go Science Crazy High Form Porcelain Crucibles with Lids
Constructed with uniform thickness and density for stable mechanical strength. GSC International High Form Porcelain Crucibles with Lids can withstand temperatures of up to 1050°C.
Supplier: GSC Go Science Crazy CHF3020
Alert:
Intended for ages 13 and up.
Description
- High quality, high form porcelain crucibles
- Porcelain is uniform in thickness and dense, which allows stable mechanical strength and little permeability
- Glazed on the inside and outside surfaces, including lids
- When properly used the glazed pieces can withstand temperatures of 1050°C
Specifications
With Lid | |
Glazed interior and exterior | |
50 mL |
Porcelain | |
1050°C |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction