Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell MICROFLEX™ Integra™ N86 Nitrile Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Unique nitrile formula is stretchier than other exam weight nitrile gloves
- Added dexterity to pick up small parts
- Textured finish
- Beaded cuff
- Powder free
- Ambidextrous, non-sterile
- Manufactured in an IS0 9002 certified facility
- Ideal for standard and moderate risk examination procedures, inspection, selecting, checking parts, assembly and equipment repair
Specifications
Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600, ISO 9001 |
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
Chemotherapeutic Rated | No |
Applications | Industrial, Healthcare, Laboratory |
AQL | 1.5 |
Color | Blue |
Disposable | Yes |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 7.5 mil |
Finger Thickness (Metric) | 0.19 mm |
Show More |