Learn More
Malvern Panalytical High Enrgy Ss Venturi Dispersr
Supplier: Malvern Panalytical MAZ2071
High Energy stainless steel venturi disperser for use with the Aero S dry powder disperser. This disperser includes an impaction plate, enabling the dispersion of robust particles which are strongly agglomerated. Note: this disperser is not recommended for use with fragile particles.