Learn More
Bel-Art™ HiFlow Vacuum Aspirator Collection System Accessory, Hydrophobic Filter
Use with HiFlow Vacuum Aspirator Collection System. This compact, portable system is ideal for vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes and is available with the original strength or HiFlow pump.
Supplier: Bel-Art™ F199170255
Description
Self-contained system with 1.0 gallon bottle, tubing and vacuum pump is lightweight, compact and easy to use. It is practical whenever a house vacuum is absent or not working
- Replaceable hydrophobic filters (6/pack)
- Protects the pump motor from fluid contamination
Specifications
HiFlow Vacuum Aspirator Collection System | |
6/Pk. | |
Hydrophobic Filter |