Learn More
Cole-Parmer Extech 42545 Hi-Temperature Infrared Thermometer (50: 1)
Supplier: Cole-Parmer 9041521
With wireless USB interface, thermometer transmits temperature data to a PC. Adjustable high/low alarm alerts user visually and audibly when temperature exceeds programmed limits. Adjustable emissivity for better accuracy on different surfaces. MAX/MIN/AVG/DIF features display highest, lowest, average, and MAX minus MIN values. High Resolution of 0.1 up to 199.9. Laser pointer identifies target area.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.