Learn More
Electron Microscopy Sciences Hi-Intensity UV Lamp, 220 V
Supplier: Electron Microscopy Sciences 72415
These UV Lamps are designed to provide a beam of 365nm wave with an average 7000uW/cm2 at 15" (38.2cm). Its powerful 100-watt light source fluoresces fine particles and hard to see surface defects. The lifetime of the light bulb is 2,000 hours. At 18" the lamp produces a highly intense center of UV irradiance about 5" in diameter. This offers the microscopist a quality tool in staining and polymerization other uses. The lamp head rotates a full 360, and is mounted into a removable pistol grip handle, which inserts firmly into the transformer base ( as a stand); it weighs only 2 1/2 lbs. It comes with a 5" (12.5cm) diameter black light filter which is recessed in the head. If more details are needed in order to study the subject, a second filter is available as an option that reduces the visible light. Total cord length is 15 ft. Lifetime 2000 hr. At 18" produces 5" of UV irradiance. 5" black light filter.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.