Learn More
Transcat Eil HH REFERENCE THERMOMETER 1-CHN
Supplier:  Transcat Eil 3376516
Encompass_Preferred
The 1523 Reference Thermometer measures, graphs, and records PRTs, thermocouples, and thermistors. This thermometer readout delivers exceptional accuracy, wide measurement range, logging, and trending, all in a handheld tool you can take anywhere.The 1523 lets you handle field applications, laboratory measurements, and data logging with ease.Three sensor typesPRTs: –200 °C to 1000 °CThermocouples –200 °C to 2315 °CPrecision thermistors: –50 °C to 150 °CAccuracyPRTs: up to ±0.011 °CThermocouples: ±0.24 °C for J,K,L,MPrecision thermistors: ±0.002 °CFast modePRTs: 0.45 seconds per sampleThermocouples: 0.3 second per samplePrecision thermistors: 0.3 seconds per sampleGraphical display128x64 backlit LCD graphic displayPlot and scale trends in real timeSimultaneous dual channel readings
Catalog No. NC2395618