Learn More
Perkin Elmer US LLC HGA Tube Alignment Tool for Non-Zeeman Furnaces
Supplier: Perkin Elmer US LLC B0080112
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The tool is compatible with the following list of PerkinElmer Atomic Absorption (AA) instruments: HGA-300, HGA-400, HGA-500, HGA-600, HGA-700, HGA-800, HGA-850, HGA-900, AAnalyst 700 and PinAAcle 900H. The alignment tool was designed to be used in non-Zeeman type furnaces.