Learn More
Perkin Elmer US LLC HGA Graphite Walled Tubes, Pkg. 50
Supplier: Perkin Elmer US LLC B0091504
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Standard pyrocoated graphite walled tubes are produced in a quality controlled manner to ensure reliable temperature calibration in PerkinElmer graphite furnaces.