Learn More
Perkin Elmer US LLC HGA Graphite Tubes with L'vov Groove, Pkg. 10
Supplier: Perkin Elmer US LLC B0121092
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Standard pyrocoated graphite tubes are grooved to accept L'vov platforms and are produced in a quality controlled manner to ensure reliable temperature calibration in PerkinElmer graphite furnaces.