Learn More
Regis Technologies Inc GC Regeant, Regis, HFBA (10 x 1 GM)
SDP
Supplier:  Regis Technologies Inc 1270851200
Encompass_Preferred
HFBA: Heptafluorobutyric Anhydride is an Acylation Reagent. HFBA is frequently used for the confirmation of drugs of abuse and reacts with alcohols, amines, and phenols. Bases such as triethylamine and trimethylamine can be added to promote reactivity to HFBA. HFBA is most commonly used for ECD because its derivatives are most sensitive to ECD.
Catalog No. 50-442-933