Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HexArmor™ Rig Lizard™ 2021 Impact Protection Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- IR-X™ Impact Exoskeleton™ with high-flex design keeps hands safe from smash & impact hazards
- Reinforced index finger and thumb crotch for increased longevity
- SlipFit™ cuff adds comfort and convenience
- Durable TP-X™ palm with reinforced stitching pattern
- Hi-vis color scheme for increased visual awareness on the job site
- Launderable
Specifications
Certifications/Compliance | ANSI/ISEA 138 Level 1 Impact Protection |
Cuff Material | SlipFit™ |
Color | Hi-vis Lime Green, Black |
Cut Resistance Rating | ANSI A3 |
Hand | Specific |
Material | IR-X Impact Exoskeleton |
Product Type | Mechanic's Gloves |
Size | Small |
For Use With (Application) | General Work, Extrication, Oil/Gas Operations, Machine Repair, and Mechanics |
Puncture Rating | ASTM F2878 LEVEL 4 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction