Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Hercules™ NSR 3041 Gloves
SDP

Catalog Number p-3888710
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

ANSI/ISEA Cut Level A9 gloves feature full coverage design and pre-curved shape for comfort and protection.

  • SuperFabric™ brand material offers highest needlestick-resistance available
  • 360° degree cut protection - ANSI/ISEA Cut Level A9, Needlestick Level 5
  • Full coverage design and pre-curved shape for comfort and protection
  • Specialized silicone gripping surface on palm
  • Launderable
Coating Material Silicone
Color Black
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4X12F
For Use With (Application) Fine-toothed animal handling, wastewater and sewage treatment
Model 3041-M
Series Hercules™ NSR 3041
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit