Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES1 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA503958S
Description
HES1 Monoclonal antibody specifically detects HES1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
HES1 | |
Monoclonal | |
1.01 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86TI2 | |
Mouse | |
Affinity chromatography | |
RUO | |
91039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI1H8 | |
Unconjugated | |
DPP9 | |
BHLHB39; class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; hairy and enhancer of split 1 (Drosophila); hairy and enhancer of split 1,; hairy homolog; Hairy-like; Hairy-like protein; hes family bHLH transcription factor 1; HES1; Hes-1; hHL; HL; HRY; RHL; transcription factor HES-1 | |
Full length human recombinant protein of human DPP9 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction