Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parker HEPA Main Filter
An extremely high efficiency (99.97% at 0.3 μm) True-HEPA box filter features a gasket on the frame, and a clamp to provide sealing against by-pass.
Supplier: Parker 2971351AGS
Description
- Extremely high efficiency (99.97% at 0.3 μm) True-HEPA box filter
- Features a gasket on the frame, and a clamp to provide sealing against by-pass
- CRYSTAL-AIRE™uv system requires one HEPA filter per change out
- Replace every 36 months or when filter change light is illuminated
Specifications
0.9997 | |
CRYSTAL-AIRE™uv System | |
0.30 μm | |
36 Months |
HEPA | |
Gasket, Clamp | |
Main Filter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title