Learn More
Fine Science Tools Fine Hemostats, Stainless Steel, Curved, 12 cm
Supplier: Fine Science Tools 1300712
Tip Shape: Curved/ Tips: Smooth/ Tip Width: 1mm/ Clamping Length: 19mm/ Alloy / Material: Stainless Steel/ Length: 12cm