Learn More
New England Biolabs, Inc. Hemo KlenTaq – 1000 reactions
Supplier: New England Biolabs, Inc. M0332L
Hemo KlenTaq is a truncated version of Taq DNA Polymerase, lacking the first 280 amino acids. Hemo KlenTaq also contains mutations that make it resistant to inhibitors present in whole blood. It can amplify directly from whole blood from humans and mice. Hemo KlenTaq tolerates up to 20% whole blood in a 25 ul reaction (30% in 50 ul reaction).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.