Learn More
Perkin Elmer US LLC Hellma Traycell 10.2 mm (Factor*50) Cap
Supplier:  Perkin Elmer US LLC L7110292
Encompass_Preferred
The Hellma Traycell Accessory is a fiber-optic, ultra-micro cell designed for UV/Vis analysis of DNA/RNA and proteins. The dimensions of the TrayCell are equivalent to a standard cuvette, allowing it to work in most spectrophotometers. LAMBDA spectrometers require a beam height of 15 mm. Features and Benefits include: Suitable for almost any current spectrophotometers Ideally suited for very small measurement volumes: 0.7 L to 10 L Trouble-free measurement of the sample at different optical path lengths simply by exchanging the cap [caps with dilution factors: 5, 10, 50 and 100] Fast and simple cleaning of the optics before measuring the next sample - the TrayCell remains in the cell holder! Samples can be reused after the measurement simply by piping them off During measurements, TrayCell shows excellent shows excellent reproducibility.
Catalog No. 50-211-2086