Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scientific Cell Company Hellma Analytics Ultrapure Water Filter
Reference filter for testing to stray light according to Ph. Eur.
Supplier: Scientific Cell Company 667UV12
Description
- Certified Wavelength: 198, 200, 300, 400nm
- Cut-off slit width: 2nm
- For use with 667.100-UV, 667.101-UV Liquid Filter Set
- In high-quality storage case with DAkkS calibration certificate
Specifications
Purified Water (blank) | |
Reference filter for testing to stray light according to Ph. Eur. | |
667.100-UV, 667.101-UV Liquid Filter Set |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction