Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scientific Cell Company Hellma Analytics Sodium Iodide Filter
Liquid filter for testing stray light.
Supplier: Scientific Cell Company 667UV10
Description
- Liquid filters with sharp cut offs in transmission at specified wavelengths
- Detection of stray light in the UV range
- Certified Wavelength: 259nm
- Cut-off slit width: 2nm
- For use with 667.101-UV
- In high-quality storage case with DAkkS calibration certificate
Specifications
Sodium Iodide Filter | |
Detection of stray light in the UV range | |
667.101-UV Liquid Filter Set | |
Quartz SUPRASIL™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction