Learn More
Scientific Cell Company Hellma Analytics Potassium Dichromate in HClO Filter
Supplier:  Scientific Cell Company 667UV 100
 View more versions of this product
Catalog No. 14-397-530
Add to Cart
Description
Description
Specifications
Specifications
DIN EN ISO 17025, NIST
Potassium Dichromate in HClO Filter
100 g
235, 257, 313, 350 nm
For testing for stray light according to Ph. Eur.
Product Suggestions
Product Suggestions