Learn More
Scientific Cell Company Hellma Analytics Potassium Dichromate in HClO Filter
Supplier: Scientific Cell Company 667UV 80
Specifications
DIN EN ISO 17025, NIST | |
Potassium Dichromate in HClO Filter | |
80 g |
235, 257, 313, 350 nm | |
For testing the absorption and linearity |