Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scientific Cell Company Hellma Analytics Perchloric Acid
Reference filter for testing the photometric accuracy.
Supplier: Scientific Cell Company 667UV14
Description
- Certified Wavelength: 235, 257, 313, 350nm
- Cut-off slit width: 2nm
- For use with 667.301-UV, 667.400-UV Liquid Filter Set
- In high-quality storage case with DAkkS calibration certificate
Specifications
Perchloric Acid (blank) | |
Reference filter for testing the photometric accuracy. | |
667.301-UV, 667.400-UV Liquid Filter Set |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction