Learn More
Scientific Cell Company Hellma Analytics Liquid Filters
Supplier: Scientific Cell Company 667UV20
Specifications
DIN EN ISO 17025, NIST | |
Liquid Filter | |
Potassium dichromate in perchloric acid (0.002% K2CR2O7) |
235, 257, 313, 350 nm | |
For testing the absorption and linearity |