Learn More
Heidolph™ Stirrer Chucks for RZR Overhead Stirrer
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Accessories for use with RZR Overhead Stirrers
$412.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
13-880-151
![]() |
Heidolph™
036300570 |
RZR 2020 - 2102 Stirrers |
Each for $412.00
|
|
|||||
13-880-150
![]() |
Heidolph™
036300560 |
RZR 1 Stirrer | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
13-889-463
![]() |
Heidolph™
036306000 |
2102 Z Stirrer | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
13880150
- For use with RZR 1 stirrer
- 8 mm
- For use with RZR 2020 - 2102 stirrers
- 10.5 mm
- For use with 2102 Z stirrer
- 10.5 mm
Specifications
Stirrer Chuck |