Learn More
Heidolph™ Overhead Stirrer Impellers - Propeller Style
Hei-TORQUE Propeller Style Impellers are recommended for applications which require average or high speed and medium to high viscosity; excellent mixing for homogenization and suspensions
Supplier: Heidolph™ 036300430
Description
- Excellent mixing properties for homogenization and suspensions
- For mixing tasks with medium or high viscosity
- Axial flow
- Made from PTFE or stainless-steel
Specifications
2.6 in. | |
6.6 cm | |
Mixing tasks with medium or high viscosity, excellent mixing properties for homogenization and suspensions | |
400 mm | |
48 oz. |
0.31 in. | |
0.8 cm | |
15.74 in. | |
Propeller Impeller | |
3 lb. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov